Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177624.3 | NP_808292.1 | 4 | 391 | ||
GenBank mRNA | AK040281.1 | - | 4 | 392 | ||
BC126945.1 | - | 4 | 392 | 40090204 |