Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001436.3 | NP_001427.2 | 3, 4 | 404 | ||
XM_005258651.2 | XP_005258708.1 | 3, 4 | 382 | |||
XM_011526623.2 | XP_011524925.1 | 2, 3 | 185 | |||
GenBank mRNA | AB451384.1 | - | 2, 3 | 290 | ||
AK296950.1 | - | 3, 4 | 369 | |||
AK308649.1 | - | 3, 4 | 366 | |||
BC015218.2 | - | 1, 2 | 109 | |||
BC019260.1 | - | 3, 4 | 358 | 3504198 | ||
BC019609.1 | - | 1, 2 | 108 | |||
BG831506.1 | - | 3, 4 | 403 | |||
BT006830.1 | - | 2, 3 | 290 | |||
BT020144.1 | - | 2, 3 | 290 | |||
CR457069.1 | - | 2, 3 | 290 | |||
M59849.1 | - | 3, 4 | 318 | |||
X56597.1 | - | 3, 4 | 349 |