Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630003M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131021.2 | NP_001124493.1 | 3 | 452 | ||
NM_177657.5 | NP_808325.2 | 3 | 452 | |||
GenBank mRNA | AK052605.1 | - | 3 | 451 | ||
AK168367.1 | - | 3 | 453 | |||
BC137959.1 | - | 3 | 363 | 8860978 | ||
BC137960.1 | - | 3 | 363 | 8860979 |