Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001441.2 | NP_001432.2 | 3 | 418 | ||
GenBank mRNA | BC093632.1 | - | 3 | 360 | 7939477 | |
BC110404.1 | - | 3 | 348 | 5728192 | ||
BC111941.1 | - | 3 | 360 | 8327409 | ||
BG718115.1 | - | 3 | 425 | |||
U82535.1 | - | 3 | 371 |