Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177625.4 | NP_808293.1 | 4 | 823 | ||
GenBank mRNA | AK077874.1 | - | 3 | 1144 | ||
AK081214.1 | - | 4 | 799 |