Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctso | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177662.2 | NP_808330.1 | 3, 4 | 401 | ||
GenBank mRNA | AK034490.1 | - | 3, 4 | 401 | ||
AK049470.1 | - | 3, 4 | 374 | |||
AK143683.1 | - | 3, 4 | 417 | |||
AK165930.1 | - | 3, 4 | 363 | |||
AK166103.1 | - | 3, 4 | 370 | |||
BC044664.1 | - | 3, 4 | 293 | |||
BC098219.1 | - | 3, 4 | 375 | 1066724 |