Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177664.5 | NP_808332.3 | 1 | 800 | ||
GenBank mRNA | AF541279.1 | - | 1 | 801 | ||
AK046782.1 | - | 1 | 801 | |||
AK079635.1 | - | 1 | 134 | |||
AK129149.1 | - | 1 | 396 | |||
AY266266.1 | - | 1 | 801 | |||
AY266267.1 | - | 1 | 801 | |||
BC082323.1 | - | 1 | 330 | |||
BC137701.1 | - | 1 | 395 | 8860720 | ||
BC137702.1 | - | 1 | 396 | 8860721 |