Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177696.1 | NP_001171167.1 | 3 | 377 | ||
NM_001177697.1 | NP_001171168.1 | 3 | 377 | |||
NM_001177698.1 | NP_001171169.1 | 3 | 377 | |||
NM_001177699.1 | NP_001171170.1 | 3 | 377 | |||
NM_001177700.1 | NP_001171171.1 | 3 | 377 | |||
NM_177678.7 | NP_808346.3 | 3 | 377 | |||
GenBank mRNA | AJ748602.1 | - | 3 | 333 | ||
AJ748603.1 | - | 3 | 333 | |||
AK046243.1 | - | 3 | 328 | |||
AK046879.1 | - | 3 | 326 | |||
AK082707.1 | - | 3 | 327 | |||
AK154494.1 | - | 3 | 379 | |||
AY274116.1 | - | 3 | 304 | |||
BC141125.1 | - | 3 | 237 | 9055737 | ||
DQ413175.1 | - | 2 | 206 |