Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001273.3 | NP_001264.2 | 19 | 3037 | ||
NM_001297553.1 | NP_001284482.1 | 18 | 2882 | |||
XM_005253668.3 | XP_005253725.1 | 19 | 2968 | |||
XM_006718958.1 | XP_006719021.1 | 19 | 3043 | |||
XM_006718959.1 | XP_006719022.1 | 19 | 3043 | |||
XM_006718960.1 | XP_006719023.1 | 19 | 3043 | |||
XM_006718961.2 | XP_006719024.1 | 19 | 2968 | |||
XM_006718962.1 | XP_006719025.1 | 19 | 3004 | |||
XM_017018725.1 | XP_016874214.1 | 19 | 3043 | |||
XM_017018726.1 | XP_016874215.1 | 19 | 3043 | |||
XM_017018727.1 | XP_016874216.1 | 19 | 3043 | |||
XM_017018728.1 | XP_016874217.1 | 19 | 3043 | |||
XM_017018729.1 | XP_016874218.1 | 19 | 2968 | |||
XM_017018730.1 | XP_016874219.1 | 19 | 3004 | |||
XM_017018731.1 | XP_016874220.1 | 19 | 3004 | |||
XM_017018732.1 | XP_016874221.1 | 19 | 2929 | |||
XM_017018733.1 | XP_016874222.1 | 19 | 2929 | |||
XM_017018734.1 | XP_016874223.1 | 19 | 2929 | |||
GenBank mRNA | AK128741.1 | - | 18 | 2927 | ||
BC038596.1 | - | 19 | 2978 | 5528023 | ||
CR936680.1 | - | 18 | 2790 | |||
X86691.1 | - | 18 | 2914 |