Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164620.1 | NP_001158092.1 | 5 | 1010 | ||
NM_177616.3 | NP_808284.2 | 6 | 1146 | |||
GenBank mRNA | AK076934.1 | - | 6 | 1074 | ||
AK133165.1 | - | 5 | 1011 | |||
AK143216.1 | - | 4 | 665 | |||
AK220409.1 | - | 6 | 1127 | |||
BC120737.1 | - | 3 | 508 | 40129660 | ||
BC120739.1 | - | 3 | 508 | 40129662 |