Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931409K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177676.6 | NP_808344.3 | 18, 19 | 2465 | ||
GenBank mRNA | AK029839.1 | - | 17, 18 | 2897 | ||
AV271477.2 | - | 2, 3 | 411 | |||
BC137999.1 | - | 14, 15 | 2186 | 8861018 | ||
BC145792.1 | - | 14, 15 | 2188 | 40131095 |