Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177696.1 | NP_001171167.1 | 22 | 2702 | ||
NM_001177697.1 | NP_001171168.1 | 20 | 2561 | |||
NM_001177698.1 | NP_001171169.1 | 19 | 2531 | |||
NM_001177699.1 | NP_001171170.1 | 19 | 2528 | |||
NM_001177700.1 | NP_001171171.1 | 18 | 2429 | |||
NM_177678.7 | NP_808346.3 | 20 | 2546 | |||
GenBank mRNA | AJ748602.1 | - | 20 | 2502 | ||
AJ748603.1 | - | 18 | 2385 | |||
AK046243.1 | - | 20 | 2497 | |||
AK046879.1 | - | 20 | 2512 | |||
AK082707.1 | - | 20 | 2533 | |||
AK154494.1 | - | 19 | 2533 | |||
AY274116.1 | - | 19 | 2454 | |||
BC141125.1 | - | 19 | 2388 | 9055737 |