Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177687.3 | NP_808355.1 | 4, 5 | 635 | ||
GenBank mRNA | AK045467.1 | - | 4, 5 | 636 | ||
AK206151.1 | - | 2, 3 | 139 | |||
BC109358.1 | - | 4, 5 | 507 | 40057562 | ||
BC109359.1 | - | 4, 5 | 523 | 40057564 |