Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177696.3 | NP_808364.2 | 18 | 2167 | ||
GenBank mRNA | AK029527.1 | - | 18 | 2171 | ||
AK161500.1 | - | 18 | 2134 | |||
BC140990.1 | - | 16 | 1895 | 9055602 |