Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177716.3 | NP_808384.2 | 1 | 935 | ||
GenBank mRNA | AK030707.1 | - | 1 | 480 | ||
AK080988.1 | - | 1 | 1081 | |||
BC090621.1 | - | 1 | 845 | 6828730 | ||
BC095968.1 | - | 1 | 415 | 6492111 |