Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177759.3 | NP_808427.1 | 2 | 592 | ||
GenBank mRNA | AK082047.1 | - | 2 | 593 | ||
BC075662.1 | - | 2 | 571 | |||
BC150892.1 | - | 2 | 382 | 9087803 | ||
BC172040.1 | - | 2 | 380 | 9054326 |