Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300965.1 | NP_001287894.1 | 5, 6 | 592 | ||
NM_001321.2 | NP_001312.1 | 5, 6 | 617 | |||
GenBank mRNA | AB209321.1 | - | 2, 3 | 4769 | ||
AY349166.1 | - | 5, 6 | 553 | |||
BC000992.2 | - | 5, 6 | 565 | 3448693 | ||
BT019913.1 | - | 4, 5 | 513 | |||
CN408330.1 | - | 5, 6 | 563 | |||
U46006.1 | - | 4, 5 | 513 | |||
U57646.1 | - | 5, 6 | 569 |