Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177752.4 | NP_808420.1 | 2 | 215 | ||
GenBank mRNA | AK078516.1 | - | 2 | 216 | ||
BC089459.1 | - | 2 | 190 | 30630337 |