Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193416.2 | NP_001180345.1 | 17 | 2235 | ||
NM_001193417.2 | NP_001180346.1 | 16 | 2190 | |||
NM_001356.4 | NP_001347.3 | 17 | 3092 | |||
NR_126093.1 | 17 | 3092 | ||||
XM_011543892.2 | XP_011542194.1 | 16 | 2129 | |||
XM_017029313.1 | XP_016884802.1 | 17 | 4437 | |||
XM_017029314.1 | XP_016884803.1 | 17 | 4434 | |||
GenBank mRNA | AF000982.1 | - | 17 | 3003 | ||
AF000983.1 | - | 17 | 3003 | |||
AK291153.1 | - | 17 | 2238 | |||
AK296906.1 | - | 6 | 669 | |||
AK297159.1 | - | 15 | 1848 | |||
AK304689.1 | - | 16 | 2190 | |||
BC011819.2 | - | 17 | 2217 | 3617040 | ||
U50553.1 | - | 17 | 2456 |