Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177763.3 | NP_808431.2 | 4 | 2405 | ||
GenBank mRNA | AK046437.1 | - | 5 | 2313 | ||
AK220449.1 | - | 5 | 2257 |