Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177781.4 | NP_808449.1 | 13 | 1631 | ||
GenBank mRNA | AK045771.1 | - | 13 | 1632 | ||
AY231177.1 | - | 13 | 1604 | |||
BC120563.1 | - | 13 | 1622 | 40129486 | ||
BC131963.1 | - | 13 | 1622 | 40130240 | ||
KF460461.1 | - | 13 | 1604 |