Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177821.6 | NP_808489.4 | 2 | 811 | ||
GenBank mRNA | AK042627.1 | - | 2 | 807 | ||
BC144976.1 | - | 2 | 665 | 9053524 | ||
BC150681.1 | - | 2 | 665 | 9087592 |