Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933431E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030251.1 | - | 4 | 2205 | ||
AK036227.1 | - | 3 | 2156 | |||
AK147824.1 | - | 4 | 2180 | |||
BC060136.1 | - | 3 | 2583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177858.1 | - | 2156 | Chr. - - [-] on Build |