Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177621.1 | NP_001171092.1 | 8 | 1356 | ||
NM_001177622.1 | NP_001171093.1 | 8 | 1138 | |||
NM_177870.5 | NP_808538.1 | 7 | 1245 | |||
GenBank mRNA | AK088682.1 | - | 8 | 1044 | ||
AK158896.1 | - | 8 | 1357 | |||
AK161633.1 | - | 7 | 1169 | |||
AY572835.1 | - | 5 | 596 | |||
BC065061.1 | - | 8 | 1138 | |||
BC085132.1 | - | 7 | 941 | 6416422 | ||
BC117852.1 | - | 7 | 898 | 40094012 | ||
BC117853.1 | - | 7 | 898 | 40094013 |