Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177906.4 | NP_808574.2 | 2, 3 | 271 | ||
GenBank mRNA | AK028345.1 | - | 1, 2 | 239 | ||
AK158538.1 | - | 2, 3 | 593 | |||
BC076581.1 | - | 2, 3 | 271 | 30542711 |