Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177910.3 | NP_808578.1 | 2 | 421 | ||
GenBank mRNA | AK088295.1 | - | 2 | 403 | ||
AK148125.1 | - | 421 | ||||
AK158470.1 | - | 2 | 421 | |||
AK165816.1 | - | 2 | 423 | |||
BC061207.1 | - | 3 | 295 | 30249330 |