Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177920.5 | NP_808588.3 | 2 | 630 | ||
GenBank mRNA | AK050361.1 | - | 3 | 175 | ||
AK149459.1 | - | 2 | 611 | |||
AY575783.1 | - | 1 | 564 | |||
BC132100.1 | - | 1 | 625 | 40130377 |