Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289497.1 | NP_001276426.1 | 8 | 1057 | ||
NM_177834.4 | NP_808502.2 | 9 | 1182 | |||
GenBank mRNA | AK078883.1 | - | 8 | 1058 | ||
AY773477.1 | - | 9 | 1180 | |||
BC119559.1 | - | 9 | 1167 | 40057538 | ||
BC119560.1 | - | 9 | 1167 | 40057539 |