Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177850.3 | NP_808518.1 | 2 | 233 | ||
GenBank mRNA | AK033770.1 | - | 2 | 235 | ||
AY363993.1 | - | 2 | 231 | |||
AY648037.1 | - | 2 | 233 | |||
AY853179.1 | - | 2 | 218 | |||
BC118049.1 | - | 2 | 227 | 40106697 | ||
BC118504.1 | - | 2 | 227 | 40106694 |