Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177861.4 | NP_808529.2 | 2 | 302 | ||
GenBank mRNA | AK017238.1 | - | 2 | 274 | ||
AK039177.1 | - | 2 | 273 | |||
AK042835.1 | - | 2 | 279 | |||
AK045429.1 | - | 2 | 303 | |||
AK077246.1 | - | 2 | 353 | |||
AK084054.1 | - | 1083 | ||||
AK133059.1 | - | 2 | 283 |