Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025163.1 | NP_001020334.1 | 5 | 707 | ||
NM_001112805.1 | NP_001106276.1 | 4 | 611 | |||
NM_177888.4 | NP_808556.1 | 5 | 850 | |||
GenBank mRNA | AK039658.1 | - | 5 | 823 | ||
AK134974.1 | - | 5 | 736 | |||
AK163133.1 | - | 4 | 610 | |||
AK173164.1 | - | 4 | 631 | |||
BC085322.1 | - | 5 | 698 | 30739758 |