Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177922.2 | NP_808590.1 | 2, 3 | 269 | ||
GenBank mRNA | BC048082.1 | - | 2, 3 | 269 | 6312082 |