Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300821.1 | NP_001287750.1 | 3 | 573 | ||
NM_001417.5 | NP_001408.2 | 3 | 573 | |||
GenBank mRNA | AB076839.1 | - | 2 | 347 | ||
AK222528.1 | - | 3 | 375 | |||
AK293731.1 | - | 3 | 375 | |||
AK299329.1 | - | 3 | 375 | |||
AK299526.1 | - | 3 | 376 | |||
AK310244.1 | - | 4 | 422 | |||
BC073139.1 | - | 3 | 363 | 6527540 | ||
BC073154.1 | - | 3 | 363 | 6015183 | ||
BC098437.1 | - | 3 | 357 | 6459393 | ||
BP236621.1 | - | 3 | 392 | |||
CR456958.1 | - | 2 | 347 |