Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177909.5 | NP_808577.3 | 13, 14 | 1699 | ||
GenBank mRNA | AK037058.1 | - | 13, 14 | 1683 | ||
AK156494.1 | - | 13, 14 | 1660 | |||
AK172687.1 | - | 13, 14 | 1671 | |||
BC099942.1 | - | 13, 14 | 1673 | 30103308 | ||
BC153825.1 | - | 13, 14 | 1677 | 40093280 |