Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177910.3 | NP_808578.1 | 9 | 1529 | ||
GenBank mRNA | AK088295.1 | - | 9 | 1511 | ||
AK158470.1 | - | 9 | 1529 | |||
AK165816.1 | - | 8 | 1614 | |||
AK205906.1 | - | 1 | 17 | |||
BC061207.1 | - | 10 | 1403 | 30249330 |