Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025662.5 | NP_079938.1 | 1, 2 | 136 | ||
NM_178016.3 | NP_821135.1 | 1, 2 | 136 | |||
GenBank mRNA | AK013853.1 | - | 1, 2 | 133 | ||
AK043033.1 | - | 125 | ||||
AK046246.1 | - | 1, 2 | 124 | |||
AK077893.1 | - | 1, 2 | 137 | |||
AK082806.1 | - | 1, 2 | 129 | |||
AK160801.1 | - | 1, 2 | 131 | |||
AK168666.1 | - | 118 | ||||
BC023256.1 | - | 1, 2 | 118 | |||
BC060175.1 | - | 1, 2 | 122 | 6827521 |