Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 1 | 571 | ||
GenBank mRNA | AK010160.1 | - | 1 | 507 | ||
AK031589.1 | - | 1 | 574 | |||
AK035321.1 | - | 1 | 524 | |||
AK150929.1 | - | 1 | 492 | |||
AK155578.1 | - | 1 | 532 | |||
AK165539.1 | - | 1 | 510 | |||
AK171154.1 | - | 1 | 530 | |||
AK185841.1 | - | 1 | 25 | |||
AK193143.1 | - | 1 | 25 | |||
AK194762.1 | - | 1 | 25 | |||
AK214365.1 | - | 1 | 21 | |||
BC049180.1 | - | 1 | 534 | 6314347 | ||
BC052721.1 | - | 1 | 495 | 6809853 | ||
BC062972.1 | - | 1 | 495 | 6809878 |