Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK196234.1 | - | 13 |
Gene Symbol: Trp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178111.3 | NP_835212.1 | 2, 3 | 344 | ||
GenBank mRNA | AK146589.1 | - | 2, 3 | 349 | ||
AK170132.1 | - | 2, 3 | 382 | |||
BC036958.1 | - | 1, 2 | 151 | 4504446 | ||
BC043086.1 | - | 2, 3 | 344 | 6417598 |