Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114182.2 | NP_001107654.1 | 13 | 1952 | ||
NM_001145256.1 | NP_001138728.1 | 12 | 1806 | |||
NM_001145257.1 | NP_001138729.1 | 11 | 1758 | |||
NM_001145258.1 | NP_001138730.1 | 10 | 1588 | |||
NM_016123.3 | NP_057207.2 | 12 | 1904 | |||
XM_005268943.3 | XP_005269000.1 | 12 | 2009 | |||
XM_005268944.4 | XP_005269001.1 | 12 | 1973 | |||
XM_005268945.4 | XP_005269002.1 | 12 | 2217 | |||
XM_005268947.4 | XP_005269004.1 | 13 | 1963 | |||
XM_005268948.2 | XP_005269005.1 | 14 | 2003 | |||
XM_005268949.2 | XP_005269006.1 | 11 | 1863 | |||
XM_006719438.3 | XP_006719501.1 | 13 | 2265 | |||
XM_006719439.2 | XP_006719502.1 | 12 | 2370 | |||
XM_006719440.2 | XP_006719503.1 | 12 | 1781 | |||
XM_006719441.2 | XP_006719504.1 | 11 | 1666 | |||
XM_006719442.2 | XP_006719505.1 | 12 | 1670 | |||
XM_006719443.3 | XP_006719506.1 | 11 | 1813 | |||
XM_011538431.2 | XP_011536733.1 | 13 | 2021 | |||
XM_011538432.1 | XP_011536734.1 | 13 | 2057 | |||
XM_011538433.2 | XP_011536735.1 | 12 | 2636 | |||
XM_017019390.1 | XP_016874879.1 | 14 | 2138 | |||
XM_017019391.1 | XP_016874880.1 | 13 | 2103 | |||
GenBank mRNA | AF155118.1 | - | 12 | 1871 | ||
AK000528.1 | - | 12 | 1851 |