Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178387.3 | NP_848474.2 | 7, 8 | 1231 | ||
GenBank mRNA | AK006916.1 | - | 6, 7 | 909 | ||
BC050799.1 | - | 6, 7 | 1133 | |||
BC120503.1 | - | 4, 5 | 683 | |||
BC120505.1 | - | 4, 5 | 683 | |||
BC144721.1 | - | 4, 5 | 683 | |||
BC144723.1 | - | 4, 5 | 683 |