Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001440.3 | NP_001431.1 | 7 | 4156 | ||
NR_073468.1 | 5 | 1439 | ||||
XM_011544440.2 | XP_011542742.1 | 6 | 3932 | |||
GenBank mRNA | AB007042.1 | - | 7 | 3993 | ||
AB011091.1 | - | 7 | 4015 | |||
AF001690.1 | - | 7 | 4001 | |||
AF029231.1 | - | 5 | 3662 | |||
AF083551.1 | - | 7 | 3993 | |||
AK124619.1 | - | 1 | 1316 | |||
CR749646.1 | - | 2 | 4163 |