Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178599.4 | NP_848714.1 | 2 | 167 | ||
GenBank mRNA | AK009275.1 | - | 2 | 156 | ||
AK012653.1 | - | 2 | 153 | |||
AK013015.1 | - | 2 | 138 | |||
AK013371.1 | - | 2 | 140 | |||
AK017777.1 | - | 2 | 82 | |||
AK077934.1 | - | 2 | 167 | |||
AK151560.1 | - | 2 | 150 | |||
AK158078.1 | - | 2 | 150 | |||
AK170011.1 | - | 2 | 140 | |||
AK181273.1 | - | 2 | 79 | |||
AK212240.1 | - | 1 | 66 | |||
BC098183.1 | - | 2 | 143 | 5150466 |