Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178603.4 | NP_848718.1 | 2 | 1793 | ||
GenBank mRNA | AK002646.1 | - | 2 | 1781 | ||
AK007405.1 | - | 2 | 1790 | |||
AK031229.1 | - | 2 | 1794 | |||
AK049421.1 | - | 2 | 1783 | |||
BC021318.1 | - | 2 | 1800 | 5035369 |