Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010404.3 | NP_034534.1 | 1 | 369 | ||
NM_177981.2 | NP_817090.1 | 1 | 369 | |||
GenBank mRNA | AJ000262.1 | - | 1 | 315 | ||
AJ002271.1 | - | 1 | 315 | |||
AK138436.1 | - | 315 | ||||
BC034089.1 | - | 1 | 307 | 4481735 | ||
BC053043.1 | - | 1 | 356 | 5694628 | ||
BY288360.1 | - | 1 | 370 |