Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178028.2 | NP_821171.1 | 5 | 514 | ||
GenBank mRNA | AY163546.1 | - | 5 | 514 |