Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133829.2 | NP_598590.2 | 2 | 1057 | ||
NM_178081.4 | NP_835182.1 | 2 | 1057 | |||
GenBank mRNA | AB247936.1 | - | 1 | 672 | ||
AK035995.1 | - | 2 | 1058 | |||
AK046866.1 | - | 2 | 1201 | |||
AK171946.1 | - | 2 | 1098 | |||
BC017534.1 | - | 1 | 166 | 4488390 | ||
BC029324.1 | - | 1 | 414 |