Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082476.2 | NP_001075945.1 | 2 | 261 | ||
NM_001252541.1 | NP_001239470.1 | 2 | 261 | |||
NM_001252542.1 | NP_001239471.1 | 2 | 328 | |||
GenBank mRNA | AK019617.1 | - | 2 | 258 | ||
AK143124.1 | - | 2 | 301 | |||
AK163207.1 | - | 2 | 231 | |||
AK169885.1 | - | 2 | 320 | |||
AK186395.1 | - | 44 | ||||
AK211498.1 | - | 2 | 179 | |||
BC049789.1 | - | 2 | 311 | 6331033 | ||
BQ771207.1 | - | 2 | 228 | |||
CD354207.1 | - | 652 |