Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178027.4 | NP_821170.1 | 6 | 3006 | ||
GenBank mRNA | AK007456.1 | - | 1 | 514 | ||
AK031589.1 | - | 6 | 3008 | |||
AK035321.1 | - | 6 | 2958 | |||
AK150929.1 | - | 6 | 2927 | |||
AK155578.1 | - | 6 | 2966 | |||
AK163860.1 | - | 1 | 709 | |||
AK171154.1 | - | 6 | 2965 | |||
AK216567.1 | - | 1 | 59 | |||
AK217669.1 | - | 1 | 59 | |||
AK218652.1 | - | 1 | 59 | |||
BC049180.1 | - | 6 | 2969 | 6314347 | ||
BC052721.1 | - | 6 | 2930 | 6809853 | ||
BC062972.1 | - | 6 | 2930 | 6809878 | ||
BC063783.1 | - | 2 | 1718 |