Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159595.1 | NP_001153067.1 | 10, 11 | 1417 | ||
NM_178112.5 | NP_835213.3 | 10, 11 | 1417 | |||
GenBank mRNA | AK049513.1 | - | 10, 11 | 1388 | ||
AK140829.1 | - | 10, 11 | 1412 | |||
AK169552.1 | - | 10, 11 | 1386 | |||
BC049092.1 | - | 10, 11 | 1347 | 6402150 | ||
BC050192.1 | - | 1, 2 | 131 | 30018264 | ||
BC057618.1 | - | 10, 11 | 1347 | 6402150 | ||
BC060149.1 | - | 2, 3 | 199 |