Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001460.4 | NP_001451.2 | 2 | 281 | ||
GenBank mRNA | AK312877.1 | - | 2 | 187 | ||
AL833218.1 | - | 2 | 170 | |||
BC005894.1 | - | 2 | 187 | 4293031 | ||
BI766311.1 | - | 2 | 285 | |||
BT006979.1 | - | 1 | 123 | |||
DB236882.1 | - | 2 | 244 | |||
Y09267.1 | - | 2 | 183 |